Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057538.hg.1 |
TC01003751.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057538.hg.1 |
ENST00000264531.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057538.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057538.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057539.hg.1 |
TC01003751.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057539.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057539.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017557.hg.1 |
ENST00000452067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057537.hg.1 |
TC01003751.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057537.hg.1 |
ENST00000264531.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057537.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057537.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017557.hg.1 |
ENST00000452067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229382.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229382.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229382.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229382.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229382.hg.1 |
TC0100017075.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100229385.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229385.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229385.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229385.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229385.hg.1 |
TC0100017075.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
ENST00000464938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017075.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229384.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229384.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229384.hg.1 |
ENST00000367142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229384.hg.1 |
NM_022731.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229384.hg.1 |
TC0100017075.hg.1 |
Clariom D TC |