Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022623.hg.1 |
TC09001750.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022623.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022623.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022622.hg.1 |
TC09001750.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022622.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022622.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022621.hg.1 |
TC09001750.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022621.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022621.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000457302.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000457302.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186930.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186930.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186930.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186930.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186930.hg.1 |
TC0900012290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012290.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186927.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186927.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186927.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186927.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186927.hg.1 |
TC0900012290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900186929.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186929.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186929.hg.1 |
ENST00000314412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186929.hg.1 |
NM_004479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186929.hg.1 |
TC0900012290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000457302.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000457302.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
ENST00000623196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009255.hg.1 |
NM_207511.2 |
Refseq |