Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021911.2 | NP_068711.1 | 10 - 11 | 1406 | 108 | |
GenBank mRNA | S77553.1 | - | 9 - 10 | 1188 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Homo sapiens Inventoried Probe spans exons |