Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061231.hg.1 |
TC01004059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061231.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061231.hg.1 |
ENST00000544618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061231.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061230.hg.1 |
TC01004059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061230.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061230.hg.1 |
ENST00000544618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061230.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061229.hg.1 |
TC01004059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061229.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061229.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235914.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235914.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235914.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235914.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235914.hg.1 |
TC0100018068.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100235916.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235916.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235916.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235916.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235916.hg.1 |
TC0100018068.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018068.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235913.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235913.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235913.hg.1 |
ENST00000366514.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235913.hg.1 |
NM_022366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235913.hg.1 |
TC0100018068.hg.1 |
Clariom D TC |