Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL590525.1 | - | 509 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens 1700 Inventoried |