Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
TC04000431.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000316355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000354403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000395640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000497512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000502280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006102.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
ENST00000512918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012818.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000316355.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000354403.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000395640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000502280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
ENST00000509972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
NM_004354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188540.hg.1 |
TC0400012818.hg.1 |
Clariom D TC |