Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167738.1 | NP_001161210.1 | 23 - 24 | 4037 | 60 | |
NM_020443.4 | NP_065176.3 | 26 - 27 | 5377 | 60 | ||
XM_006711609.2 | XP_006711672.2 | 24 - 25 | 4012 | 60 | ||
XM_006711610.2 | XP_006711673.2 | 23 - 24 | 3988 | 60 | ||
XM_006711611.1 | XP_006711674.1 | 24 - 25 | 4128 | 60 | ||
XM_011510097.1 | XP_011508399.1 | 25 - 26 | 5047 | 60 | ||
XM_011510098.1 | XP_011508400.1 | 25 - 26 | 5032 | 60 | ||
XM_011510099.1 | XP_011508401.1 | 24 - 25 | 5023 | 60 | ||
XM_011510100.1 | XP_011508402.1 | 25 - 26 | 4885 | 60 | ||
XM_011510101.1 | XP_011508403.1 | 24 - 25 | 4861 | 60 | ||
XM_011510102.1 | XP_011508404.1 | 23 - 24 | 4852 | 60 | ||
XM_017002751.1 | XP_016858240.1 | 24 - 25 | 4876 | 60 | ||
XM_017002752.1 | XP_016858241.1 | 22 - 23 | 3979 | 60 | ||
GenBank mRNA | AB032977.3 | - | 26 - 27 | 4710 | 60 | |
AJ488101.1 | - | 26 - 27 | 4908 | 60 | ||
AK022622.1 | - | 4 - 5 | 642 | 60 | ||
AK024265.1 | - | 11 - 12 | 1542 | 60 | ||
AY043013.1 | - | 26 - 27 | 5377 | 60 | ||
BC007523.1 | - | 12 - 13 | 1659 | 60 | ||
BX648460.1 | - | 23 - 24 | 4004 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |