Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
ENST00000489892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007511.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000307859.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000376724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
ENST00000396581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
NM_001286633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600150805.hg.1 |
TC0600007511.hg.1 |
Clariom D TC |