Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056781.hg.1 |
TC01003699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056781.hg.1 |
ENST00000272217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056781.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056781.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056781.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056784.hg.1 |
TC01003699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056784.hg.1 |
ENST00000272217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056784.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056784.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056785.hg.1 |
TC01003699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056785.hg.1 |
ENST00000272217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056785.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056785.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056783.hg.1 |
TC01003699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056783.hg.1 |
ENST00000272217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056783.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056783.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228292.hg.1 |
TC0100016944.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228294.hg.1 |
TC0100016944.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000491358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016944.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000486225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228289.hg.1 |
TC0100016944.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228293.hg.1 |
TC0100016944.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
ENST00000272217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
ENST00000614750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
NM_001256129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
NM_138795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228291.hg.1 |
TC0100016944.hg.1 |
Clariom D TC |