Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025828.hg.1 |
TC02001611.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025828.hg.1 |
ENST00000272223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025828.hg.1 |
ENST00000536433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025828.hg.1 |
NM_145260.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
ENST00000272223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
ENST00000487581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
NM_145260.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
ENST00000272223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
ENST00000487581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
NM_145260.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186266.hg.1 |
TC0200011878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
ENST00000272223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
ENST00000487581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
NM_145260.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
ENST00000272223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
ENST00000487581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
NM_145260.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186265.hg.1 |
TC0200011878.hg.1 |
Clariom D TC |