Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001292034.2 | NP_001278963.1 | 5 - 6 | 2035 | 124 | |
NM_001292035.2 | NP_001278964.1 | 5 - 6 | 2070 | 124 | ||
NM_015093.5 | NP_055908.1 | 7 - 8 | 2278 | 124 | ||
XM_011535633.2 | XP_011533935.1 | 6 - 7 | 2717 | 124 | ||
XM_017010591.1 | XP_016866080.1 | 6 - 7 | 2248 | 124 | ||
XM_017010592.1 | XP_016866081.1 | 6 - 7 | 2785 | 124 | ||
GenBank mRNA | AB018276.2 | - | 5 - 6 | 2032 | 124 | |
AF241230.1 | - | 4 - 5 | 1937 | 124 | ||
AK025538.1 | - | 3 - 4 | 398 | 124 | ||
AK295750.1 | - | 4 - 5 | 1891 | 124 | ||
AL117407.1 | - | 6 - 7 | 2177 | 124 | ||
BC035910.2 | - | 5 - 6 | 1993 | 5492359 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |