Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020051.hg.1 |
TC08001525.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020051.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020051.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
ENST00000518838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011462.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178015.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178015.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178015.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178015.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178015.hg.1 |
TC0800011462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178014.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178014.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178014.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178014.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178014.hg.1 |
TC0800011462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178016.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178016.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178016.hg.1 |
ENST00000297459.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178016.hg.1 |
NM_153015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178016.hg.1 |
TC0800011462.hg.1 |
Clariom D TC |