Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002146.hg.1 |
TC03000102.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002146.hg.1 |
ENST00000285083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002146.hg.1 |
ENST00000544043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002146.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002148.hg.1 |
TC03000102.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002148.hg.1 |
ENST00000285083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002148.hg.1 |
ENST00000544043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002148.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000486267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013795.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203561.hg.1 |
TC0300013795.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000285083.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000435829.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000442255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000452581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000605932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
ENST00000627468.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
NM_138381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203565.hg.1 |
TC0300013795.hg.1 |
Clariom D TC |