Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037040.hg.1 |
TC01002401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037040.hg.1 |
ENST00000263980.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037040.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037040.hg.1 |
ENST00000545949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037040.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000490329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
ENST00000490329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037039.hg.1 |
TC01002401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037039.hg.1 |
ENST00000263980.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037039.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037039.hg.1 |
ENST00000545949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037039.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374084.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000374089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000447808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000490329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
ENST00000490329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013417.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
ENST00000263980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
ENST00000374086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
NM_003047.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
NR_046474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199674.hg.1 |
TC0100013417.hg.1 |
Clariom D TC |