Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000878.3 | NP_000869.1 | 8 - 9 | 949 | 90 | |
GenBank mRNA | AK312860.1 | - | 8 - 9 | 943 | 90 | |
BC025691.1 | - | 8 - 9 | 932 | 5207833 | 90 | |
CR456506.1 | - | 7 - 8 | 837 | 90 | ||
M26062.1 | - | 8 - 9 | 949 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |