Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015130.2 | NP_055945.2 | 19 - 20 | 3240 | 97 | |
GenBank mRNA | AB020689.2 | - | 19 - 20 | 3043 | 97 | |
AB449892.1 | - | 19 - 20 | 2968 | 97 | ||
AK127020.1 | - | 16 - 17 | 2528 | 97 | ||
BC038804.1 | - | 10 - 11 | 1354 | 97 | ||
BC146758.1 | - | 19 - 20 | 3043 | 9007196 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |