Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010939.hg.1 |
TC03000607.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010939.hg.1 |
ENST00000264245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010939.hg.1 |
ENST00000543280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010939.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160299.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160299.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160299.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160299.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160299.hg.1 |
TC0300008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000264245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
ENST00000482743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008467.hg.1 |
NM_020754.3 |
Refseq |