Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_001536.2 | 2 - 3 | 288 | 68 | ||
GenBank mRNA | DB449030.1 | - | 2 - 3 | 288 | 68 |
Gene Symbol: TTTY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003590.1 | 2 - 3 | 288 | 68 |