Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
TC01002476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
ENST00000373470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
ENST00000373471.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038267.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000465346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000465346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000483143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000483143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000465346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000465346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000481487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000483143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000483143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
ENST00000497068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013632.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
ENST00000373471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
ENST00000496770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_001171940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_001171941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
NM_153756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100201398.hg.1 |
TC0100013632.hg.1 |
Clariom D TC |