Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
TC0X001207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
ENST00000255531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
ENST00000373034.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
ENST00000420881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
NM_001105243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
NM_001184880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016194.hg.1 |
NM_020766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000255531.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000373034.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000420881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_001105243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_001184880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_020766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000255531.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000373034.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
ENST00000420881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_001105243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_001184880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
NM_020766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170148.hg.1 |
TC0X00010326.hg.1 |
Clariom D TC |