Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000558349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000558349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000623053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
ENST00000623053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013761.hg.1 |
ENST00000389286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013761.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013761.hg.1 |
TC15001469.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500170135.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170135.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170135.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170135.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170135.hg.1 |
TC1500009505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000389286.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000558349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000558349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000559842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000561465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000623053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
ENST00000623053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009505.hg.1 |
NM_173814.5 |
Refseq |