Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016139.hg.1 |
TC01001015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016139.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016139.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016141.hg.1 |
TC01001015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016141.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016141.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016140.hg.1 |
TC01001015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016140.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016140.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168615.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168615.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168615.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168615.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168615.hg.1 |
TC0100009535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168614.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168614.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168614.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168614.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168614.hg.1 |
TC0100009535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
ENST00000464946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009535.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168613.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168613.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168613.hg.1 |
ENST00000369500.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168613.hg.1 |
NM_152367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168613.hg.1 |
TC0100009535.hg.1 |
Clariom D TC |