Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010724.hg.1 |
TC01000661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010724.hg.1 |
ENST00000234827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010724.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010724.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010725.hg.1 |
TC01000661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010725.hg.1 |
ENST00000234827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010725.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010725.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000391366.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
ENST00000585250.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_039942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010729.hg.1 |
TC01000661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010729.hg.1 |
ENST00000234827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010729.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010729.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010729.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159637.hg.1 |
TC0100008375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159642.hg.1 |
TC0100008375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159636.hg.1 |
TC0100008375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000391366.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
ENST00000585250.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_039942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008375.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000234827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000371331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
ENST00000498272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
NM_153035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
NR_130900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159641.hg.1 |
TC0100008375.hg.1 |
Clariom D TC |