Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
ENST00000324616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
ENST00000398716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
ENST00000405531.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
ENST00000468200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003672.hg.1 |
TC12000307.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007349.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000324616.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000405531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
ENST00000468200.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
NM_001031748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200151420.hg.1 |
TC1200007349.hg.1 |
Clariom D TC |