Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009675.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168714.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168714.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168714.hg.1 |
ENST00000307271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700168714.hg.1 |
NM_175571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700168714.hg.1 |
TC0700009675.hg.1 |
Clariom D TC |