Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000020.hg.1 |
ENST00000325113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000020.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000020.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000020.hg.1 |
TC11000003.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000342593.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006437.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144067.hg.1 |
TC1100006437.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000325113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000525282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
ENST00000531679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
NM_001286136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
NM_053280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144065.hg.1 |
TC1100006437.hg.1 |
Clariom D TC |