Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022493.hg.1 |
TC06001854.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022493.hg.1 |
ENST00000370570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022493.hg.1 |
ENST00000370577.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022493.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012240.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000370570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000370577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
ENST00000472827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
NM_018368.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184242.hg.1 |
TC0600012240.hg.1 |
Clariom D TC |