Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000763.hg.1 |
TC02000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000763.hg.1 |
ENST00000381786.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000763.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000765.hg.1 |
TC02000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000765.hg.1 |
ENST00000381786.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000765.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000764.hg.1 |
TC02000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000764.hg.1 |
ENST00000381786.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000764.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145764.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145764.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145764.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145764.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145764.hg.1 |
TC0200006679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145769.hg.1 |
TC0200006679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145766.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145766.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145766.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145766.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145766.hg.1 |
TC0200006679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145767.hg.1 |
TC0200006679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000381786.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
ENST00000619640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006679.hg.1 |
NM_182626.3 |
Refseq |