Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010905.hg.1 |
TC0X002306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010905.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010905.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001289.hg.1 |
TC0Y000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001289.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001289.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001290.hg.1 |
TC0Y000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001290.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010904.hg.1 |
TC0X002306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010904.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010904.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001284.hg.1 |
TC0Y000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001284.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001284.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010909.hg.1 |
TC0X002306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010909.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010909.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010910.hg.1 |
TC0X002306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010910.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001285.hg.1 |
TC0Y000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001285.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001285.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001286.hg.1 |
TC0Y000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001286.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001286.hg.1 |
ENST00000460672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001286.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010906.hg.1 |
TC0X002306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010906.hg.1 |
ENST00000381297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010906.hg.1 |
ENST00000460672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X010906.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000460672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006887.hg.1 |
ENSTR0000460672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161261.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161261.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161261.hg.1 |
TC0X00008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006887.hg.1 |
ENSTR0000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161256.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161256.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161256.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161256.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161256.hg.1 |
TC0X00008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00161260.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161260.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161260.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161260.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161260.hg.1 |
TC0X00008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161257.hg.1 |
TC0X00008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00161255.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161255.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161255.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161255.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161255.hg.1 |
TC0X00008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000381297.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
ENST00000460672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008911.hg.1 |
NM_178129.4 |
Refseq |