Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
ENST00000618933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017697.hg.1 |
ENST00000219368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017697.hg.1 |
ENST00000544337.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017697.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017697.hg.1 |
TC16001271.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184404.hg.1 |
TC1600010861.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000219368.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000562145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000567683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000569949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
ENST00000618933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010861.hg.1 |
NM_024306.4 |
Refseq |