Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307983.1 | NP_001294912.1 | 4 - 5 | 376 | 87 | |
XM_005263588.4 | XP_005263645.1 | 3 - 4 | 191 | 87 | ||
XM_011510592.2 | XP_011508894.1 | 4 - 5 | 677 | 87 | ||
XM_017003325.1 | XP_016858814.1 | 4 - 5 | 674 | 87 | ||
XM_017003326.1 | XP_016858815.1 | 4 - 5 | 1324 | 87 | ||
GenBank mRNA | BC016018.1 | - | 4 - 5 | 378 | 4714682 | 87 |
KU178859.1 | - | 1 - 2 | 78 | 87 | ||
KU178860.1 | - | 1 - 2 | 78 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |