Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006190.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006190.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006190.hg.1 |
TC20000457.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
ENST00000488949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007895.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154758.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154758.hg.1 |
NM_020673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000154758.hg.1 |
TC2000007895.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000154758.hg.1 |
ENST00000244040.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000154758.hg.1 |
NM_020673.2 |
Refseq |