Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |