Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001538.hg.1 |
ENST00000280481.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001538.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001538.hg.1 |
TC13000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
ENST00000482551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006919.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147021.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147021.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147021.hg.1 |
ENST00000280481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147021.hg.1 |
NM_207361.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147021.hg.1 |
TC1300006919.hg.1 |
Clariom D TC |