Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205291.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205291.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205291.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205291.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205291.hg.1 |
TC1700012355.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000331336.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
ENST00000396267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_001304947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012355.hg.1 |
NM_213597.3 |
Refseq |