Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022710.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022710.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022710.hg.1 |
TC17001687.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000503118.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000503118.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189395.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189395.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189395.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189395.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189395.hg.1 |
TC1700011097.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008395.hg.1 |
ENST00000402043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000225972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000503118.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000503118.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
ENST00000576448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011097.hg.1 |
NM_018509.3 |
Refseq |