Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002373.hg.1 |
ENST00000300403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002373.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002373.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002373.hg.1 |
TC20000198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
TC2000007089.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148380.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000300403.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
ENST00000340513.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007089.hg.1 |
NM_012112.4 |
Refseq |