Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK025304.1 | - | 1 | 1106 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |