Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
TC03001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
ENST00000341848.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
ENST00000468189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
ENST00000476844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
ENST00000492590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027520.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185049.hg.1 |
TC0300011324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000341848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000465330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000466788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000468189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000476844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000488467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000490952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
ENST00000492590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_001166243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011324.hg.1 |
NM_002012.3 |
Refseq |