Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005703.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005703.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005703.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005700.hg.1 |
ENST00000216044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005700.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005700.hg.1 |
TC22000304.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168816.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
TC2200009266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168813.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000216044.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000418601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000460605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000461428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000462332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000470836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000475959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000484657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
ENST00000488787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009266.hg.1 |
NM_004286.4 |
Refseq |