Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
TC01001720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
ENST00000358184.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
ENST00000361052.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
ENST00000432969.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026793.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229537.hg.1 |
TC0100017084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000358184.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000360218.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000468617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
ENST00000486757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_001910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017084.hg.1 |
NM_148964.2 |
Refseq |