Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017756.hg.1 |
ENST00000394412.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017756.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017756.hg.1 |
TC15001833.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178317.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178317.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178317.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178317.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178317.hg.1 |
TC1500010396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000394412.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000445906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
ENST00000558928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010396.hg.1 |
NM_198525.2 |
Refseq |