Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011908.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011908.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011908.hg.1 |
TC20000871.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011904.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011904.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011904.hg.1 |
TC20000871.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011905.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011905.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011905.hg.1 |
TC20000871.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011906.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011906.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011906.hg.1 |
TC20000871.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164394.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164394.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164394.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164394.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164394.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164397.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164397.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164397.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164397.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164397.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164392.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164392.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164392.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164392.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164392.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164393.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164393.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164393.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164393.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164393.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164395.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164395.hg.1 |
NM_006809.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164395.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164395.hg.1 |
ENST00000372813.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164395.hg.1 |
NM_006809.4 |
Refseq |