Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003688.hg.1 |
TC03000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003688.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003688.hg.1 |
ENST00000423219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003688.hg.1 |
ENST00000448498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003688.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003691.hg.1 |
TC03000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003691.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003691.hg.1 |
ENST00000423219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003691.hg.1 |
ENST00000448498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003691.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149498.hg.1 |
TC0300007074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000273173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000448498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000466887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
ENST00000496724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
NM_004803.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149495.hg.1 |
TC0300007074.hg.1 |
Clariom D TC |