Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012291.4 | NP_036423.4 | 23 - 24 | 5333 | 73 | |
XM_006719705.3 | XP_006719768.1 | 23 - 24 | 5400 | 73 | ||
XM_011539024.2 | XP_011537326.1 | 23 - 24 | 5361 | 73 | ||
XM_011539025.2 | XP_011537327.1 | 23 - 24 | 5372 | 73 | ||
XM_017020253.1 | XP_016875742.1 | 23 - 24 | 5451 | 73 | ||
GenBank mRNA | AY455930.1 | - | 22 - 23 | 5242 | 73 | |
D79987.1 | - | 23 - 24 | 5380 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |