Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005632.hg.1 |
TC09002903.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005632.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005632.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012155.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184371.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184371.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184371.hg.1 |
ENST00000502978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184371.hg.1 |
NM_001198812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184371.hg.1 |
TC0900012155.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012154.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184358.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184358.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184358.hg.1 |
ENST00000374879.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184358.hg.1 |
NM_003692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184358.hg.1 |
TC0900012154.hg.1 |
Clariom D TC |