Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001803.hg.1 |
TC05000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001803.hg.1 |
ENST00000303115.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001803.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001806.hg.1 |
TC05000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001806.hg.1 |
ENST00000303115.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001806.hg.1 |
ENST00000343305.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001806.hg.1 |
ENST00000506850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001806.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
ENST00000303115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
ENST00000509668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
ENST00000303115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
ENST00000509668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148308.hg.1 |
TC0500007138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000303115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000505093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000505875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000506850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000514217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
NR_120485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000303115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000505093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000505875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000506850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
ENST00000514217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
NM_002185.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
NR_120485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148311.hg.1 |
TC0500007138.hg.1 |
Clariom D TC |