Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012082.hg.1 |
ENST00000278618.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012082.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012082.hg.1 |
TC11000970.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
ENST00000534152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008955.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000278618.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000524411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000525660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
ENST00000533423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
NM_015423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166112.hg.1 |
TC1100008955.hg.1 |
Clariom D TC |