Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021418.hg.1 |
TC08002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021418.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021418.hg.1 |
ENST00000434736.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021418.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188177.hg.1 |
TC0800012468.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000414222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000434736.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
ENST00000473291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012468.hg.1 |
NM_001145095.1 |
Refseq |