Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009248.hg.1 |
ENST00000219454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009248.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009248.hg.1 |
TC16000656.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009247.hg.1 |
ENST00000219454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009247.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009247.hg.1 |
TC16000656.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164565.hg.1 |
TC1600008643.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000219454.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000567056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000568638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000613603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
ENST00000622779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282466.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_001282467.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008643.hg.1 |
NM_021197.3 |
Refseq |